Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106106.1 | NP_001099576.1 | 3 - 4 | 399 | 163 | |
GenBank mRNA | BC166841.1 | - | 387 | 9027959 | 163 | |
FQ212823.1 | - | 3 - 4 | 386 | 163 | ||
FQ219292.1 | - | 3 - 4 | 418 | 163 | ||
FQ220783.1 | - | 3 - 4 | 400 | 163 | ||
FQ223579.1 | - | 3 - 4 | 386 | 163 | ||
FQ225943.1 | - | 3 - 4 | 418 | 163 | ||
FQ231336.1 | - | 3 - 4 | 386 | 163 | ||
FQ233549.1 | - | 3 - 4 | 386 | 163 | ||
FQ234239.1 | - | 3 - 4 | 398 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |