Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108412.1 | NP_001101882.1 | 4 - 5 | 310 | 96 | |
XM_006253888.2 | XP_006253950.1 | 4 - 5 | 258 | 96 | ||
XM_006253889.2 | XP_006253951.1 | 4 - 5 | 531 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Detect Genomic DNA Rattus norvegicus Probe spans exons |