Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008891.2 | NP_001008891.2 | 1 - 2 | 149 | 145 | |
XM_006253835.2 | XP_006253897.1 | 1 - 2 | 186 | 145 | ||
GenBank mRNA | AY321345.1 | - | 123 | 145 | ||
BC098008.1 | - | 1 - 2 | 149 | 7454159 | 145 | |
FQ220080.1 | - | 1 - 2 | 117 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |