Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135169.2 | NP_001128641.1 | 5 - 6 | 415 | 143 | |
NM_134395.2 | NP_599222.2 | 4 - 5 | 628 | 143 | ||
XM_006253777.2 | XP_006253839.1 | 5 - 6 | 618 | 143 | ||
GenBank mRNA | BC158854.1 | - | 4 - 5 | 309 | 7936967 | 143 |
CV107072.1 | - | 1 - 2 | 143 | |||
FQ211234.1 | - | 5 - 6 | 405 | 143 | ||
FQ217516.1 | - | 5 - 6 | 405 | 143 | ||
FQ218209.1 | - | 5 - 6 | 411 | 143 | ||
FQ223586.1 | - | 5 - 6 | 395 | 143 | ||
FQ223742.1 | - | 5 - 6 | 405 | 143 | ||
FQ224495.1 | - | 5 - 6 | 411 | 143 | ||
FQ225707.1 | - | 5 - 6 | 385 | 143 | ||
FQ230083.1 | - | 5 - 6 | 433 | 143 | ||
FQ230323.1 | - | 5 - 6 | 414 | 143 | ||
FQ231364.1 | - | 5 - 6 | 395 | 143 | ||
FQ233821.1 | - | 5 - 6 | 403 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |