Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105743.1 | NP_001099213.1 | 10 - 11 | 1355 | 80 | |
GenBank mRNA | AB001453.1 | - | 10 - 11 | 1569 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |