Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033674.1 | NP_001028846.1 | 1 - 2 | 333 | 69 | |
NM_198765.2 | NP_942060.1 | 1 - 2 | 313 | 69 | ||
GenBank mRNA | AJ250312.1 | - | 1 - 2 | 313 | 69 | |
BC100660.1 | - | 1 - 2 | 333 | 7935996 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |