Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033674.1 | NP_001028846.1 | 2 - 3 | 550 | 102 | |
NM_198765.2 | NP_942060.1 | 2 - 3 | 530 | 102 | ||
GenBank mRNA | AJ250312.1 | - | 2 - 3 | 530 | 102 | |
BC100660.1 | - | 2 - 3 | 550 | 7935996 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |