Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106107.1 | NP_001099577.1 | 2 - 3 | 298 | 176 | |
XM_006253787.2 | XP_006253849.1 | 2 - 3 | 320 | 176 | ||
XM_006253788.2 | XP_006253850.1 | 2 - 3 | 320 | 176 | ||
GenBank mRNA | BC088167.1 | - | 2 - 3 | 281 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |