Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108406.1 | NP_001101876.1 | 13 - 14 | 1267 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |