Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001059910.4 | XP_001059910.2 | 4 - 5 | 711 | 80 | |
XM_002729144.4 | XP_002729190.1 | 4 - 5 | 1325 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |