Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005542.2 | NP_001005542.2 | 2 - 3 | 360 | 115 | |
GenBank mRNA | BC082103.1 | - | 1 - 2 | 144 | 7134619 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |