Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160263.1 | NP_001153735.1 | 2 - 3 | 133 | 63 | |
GenBank mRNA | AB081652.1 | - | 2 - 3 | 177 | 63 | |
AY228143.1 | - | 2 - 3 | 133 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |