Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108575.1 | NP_001102045.1 | 6 - 7 | 1284 | 73 | |
XM_006233825.2 | XP_006233887.1 | 6 - 7 | 1287 | 73 | ||
XM_006233826.2 | XP_006233888.1 | 6 - 7 | 1313 | 73 | ||
XM_006233830.2 | XP_006233892.1 | 6 - 7 | 1287 | 73 | ||
GenBank mRNA | BC168949.1 | - | 6 - 7 | 1232 | 7112751 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |