Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105739.1 | NP_001099209.1 | 2 - 3 | 586 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |