Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36rn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107814.1 | NP_001101284.1 | 5 - 6 | 568 | 63 | |
XM_008761586.1 | XP_008759808.1 | 3 - 4 | 1219 | 63 | ||
XM_008761587.1 | XP_008759809.1 | 4 - 5 | 862 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |