Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168549.1 | NP_001162021.1 | 14 - 15 | 4512 | 66 | |
XM_006233673.2 | XP_006233735.1 | 13 - 14 | 4447 | 66 | ||
XM_006233675.2 | XP_006233737.1 | 14 - 15 | 4480 | 66 | ||
XM_006233677.2 | XP_006233739.1 | 9 - 10 | 3392 | 66 | ||
GenBank mRNA | FQ211685.1 | - | 3 - 4 | 1171 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |