Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168549.1 | NP_001162021.1 | 7 - 8 | 1336 | 75 | |
XM_006233672.2 | XP_006233734.1 | 7 - 8 | 1304 | 75 | ||
XM_006233673.2 | XP_006233735.1 | 6 - 7 | 1271 | 75 | ||
XM_006233674.2 | XP_006233736.1 | 6 - 7 | 1271 | 75 | ||
XM_006233675.2 | XP_006233737.1 | 7 - 8 | 1304 | 75 | ||
XM_006233677.2 | XP_006233739.1 | 2 - 3 | 216 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |