Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135780.1 | NP_001129252.1 | 18 - 19 | 3228 | 72 | |
XM_006237954.2 | XP_006238016.1 | 18 - 19 | 3280 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |