Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135780.1 | NP_001129252.1 | 10 - 11 | 1569 | 156 | |
XM_006237954.2 | XP_006238016.1 | 10 - 11 | 1621 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |