Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238139.2 | XP_006238201.1 | 1 - 2 | 576 | 92 | |
XM_008763687.1 | XP_008761909.1 | 1 - 2 | 578 | 92 | ||
XM_008763688.1 | XP_008761910.1 | 2 - 3 | 102 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |