Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b5p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238129.2 | XP_006238191.1 | 23 - 24 | 3024 | 86 | |
XM_008763657.1 | XP_008761879.1 | 23 - 24 | 2988 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |