Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107955.2 | NP_001101425.2 | 4 - 5 | 396 | 56 | |
GenBank mRNA | BI282127.1 | - | 1 - 2 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |