Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108657.1 | NP_001102127.1 | 7 - 8 | 1346 | 71 | |
XM_006237901.2 | XP_006237963.1 | 8 - 9 | 1479 | 71 | ||
XM_006237903.2 | XP_006237965.1 | 7 - 8 | 1251 | 71 | ||
XM_006237904.2 | XP_006237966.1 | 7 - 8 | 1309 | 71 | ||
XM_008763548.1 | XP_008761770.1 | 5 - 6 | 2757 | 71 | ||
XM_008763549.1 | XP_008761771.1 | 7 - 8 | 6642 | 71 | ||
XM_008763550.1 | XP_008761772.1 | 8 - 9 | 1175 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |