Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238046.2 | XP_006238108.1 | 1 - 2 | 200 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |