Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013048.2 | NP_037180.1 | 3 - 4 | 556 | 76 | |
XM_008763588.1 | XP_008761810.1 | 5 - 6 | 563 | 76 | ||
XM_008763589.1 | XP_008761811.1 | 4 - 5 | 506 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |