Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgol2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271191.1 | NP_001258120.1 | 4 - 5 | 458 | 143 | |
XM_006244988.2 | XP_006245050.1 | 5 - 6 | 735 | 143 | ||
XM_006244989.2 | XP_006245051.1 | 5 - 6 | 736 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |