Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173098.1 | NP_775121.1 | 10 - 11 | 2421 | 69 | |
XM_006244738.2 | XP_006244800.1 | 10 - 11 | 2587 | 69 | ||
XM_008766988.1 | XP_008765210.1 | 10 - 11 | 2587 | 69 | ||
XM_008766989.1 | XP_008765211.1 | 11 - 12 | 2490 | 69 | ||
XM_008766990.1 | XP_008765212.1 | 11 - 12 | 2363 | 69 | ||
XM_008766991.1 | XP_008765213.1 | 10 - 11 | 1710 | 69 | ||
GenBank mRNA | M85301.1 | - | 10 - 11 | 2421 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Rattus norvegicus Probe spans exons |