Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022610.1 | NP_072132.1 | 3 - 4 | 542 | 144 | |
XM_006245038.2 | XP_006245100.1 | 5 - 6 | 791 | 144 | ||
GenBank mRNA | AB023134.1 | - | 3 - 4 | 542 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |