Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mitd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009714.1 | NP_001009714.1 | 284 | 192 | ||
XM_006244828.2 | XP_006244890.1 | 358 | 192 | |||
XM_008767036.1 | XP_008765258.1 | 358 | 192 | |||
GenBank mRNA | BC088263.1 | - | 284 | 7376620 | 192 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |