Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108213.1 | NP_001101683.1 | 9 - 10 | 1680 | 73 | |
XM_006244819.2 | XP_006244881.1 | 10 - 11 | 2412 | 73 | ||
XM_006244820.2 | XP_006244882.1 | 10 - 11 | 3032 | 73 | ||
XM_006244821.2 | XP_006244883.1 | 9 - 10 | 2211 | 73 | ||
XM_008767030.1 | XP_008765252.1 | 6 - 7 | 1219 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |