Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245063.2 | XP_006245125.1 | 7 - 8 | 1068 | 104 | |
XM_006245064.2 | XP_006245126.1 | 7 - 8 | 845 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |