Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hibch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013112.1 | NP_001013130.1 | 8 - 9 | 769 | 107 | |
XM_006244895.1 | XP_006244957.1 | 8 - 9 | 855 | 107 | ||
GenBank mRNA | BC083737.1 | - | 8 - 9 | 769 | 7191666 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |