Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245080.2 | XP_006245142.1 | 5 - 6 | 1962 | 66 | |
XM_008767168.1 | XP_008765390.1 | 5 - 6 | 1962 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |