Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009671.1 | NP_001009671.1 | 2 - 3 | 286 | 187 | |
XM_008767080.1 | XP_008765302.1 | 2 - 3 | 587 | 187 | ||
GenBank mRNA | BC088273.1 | - | 2 - 3 | 286 | 7380093 | 187 |
FQ219530.1 | - | 1 - 2 | 145 | 187 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |