Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106913.2 | NP_001100383.2 | 8 - 9 | 1057 | 159 | |
XM_008767083.1 | XP_008765305.1 | 7 - 8 | 986 | 159 | ||
GenBank mRNA | BC081942.1 | - | 6 - 7 | 2316 | 159 | |
BC097451.1 | - | 6 - 7 | 2273 | 159 | ||
FQ219215.1 | - | 4 - 5 | 630 | 159 | ||
FQ232068.1 | - | 8 - 9 | 1067 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |