Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138919.2 | NP_620274.1 | 1 - 2 | 225 | 111 | |
GenBank mRNA | AY017209.1 | - | 1 - 2 | 225 | 111 | |
BC061976.1 | - | 1 - 2 | 220 | 5623807 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |