Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025731.1 | NP_001020902.1 | 10 - 11 | 1515 | 101 | |
XM_006233836.2 | XP_006233898.1 | 10 - 11 | 1551 | 101 | ||
XM_006233838.2 | XP_006233900.1 | 10 - 11 | 1550 | 101 | ||
GenBank mRNA | BC099088.1 | - | 10 - 11 | 1515 | 7456179 | 101 |
FQ217346.1 | - | 10 - 11 | 1536 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |