Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022303.1 | NP_071639.1 | 9 - 10 | 1425 | 87 | |
GenBank mRNA | AF311288.1 | - | 9 - 10 | 1425 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |