Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100171.1 | NP_001093641.1 | 8 - 9 | 821 | 91 | |
XM_006233600.2 | XP_006233662.1 | 8 - 9 | 1084 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |