Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011964.1 | NP_001011964.1 | 8 - 9 | 1510 | 86 | |
XM_006233949.2 | XP_006234011.1 | 9 - 10 | 1696 | 86 | ||
XM_006233950.2 | XP_006234012.1 | 8 - 9 | 1248 | 86 | ||
XM_008761721.1 | XP_008759943.1 | 7 - 8 | 1210 | 86 | ||
XM_008761722.1 | XP_008759944.1 | 6 - 7 | 1120 | 86 | ||
XM_008761723.1 | XP_008759945.1 | 6 - 7 | 798 | 86 | ||
GenBank mRNA | BC079073.1 | - | 8 - 9 | 1510 | 7122012 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |