Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106555.1 | NP_001100025.1 | 3 - 4 | 407 | 64 | |
XM_006233581.2 | XP_006233643.1 | 3 - 4 | 792 | 64 | ||
GenBank mRNA | FQ210613.1 | - | 3 - 4 | 367 | 64 | |
FQ218819.1 | - | 3 - 4 | 367 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |