Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004231.1 | NP_001004231.1 | 5 - 6 | 703 | 95 | |
XM_006233584.2 | XP_006233646.1 | 4 - 5 | 1341 | 95 | ||
GenBank mRNA | BC079156.1 | - | 5 - 6 | 703 | 7128989 | 95 |
FQ214366.1 | - | 5 - 6 | 547 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |