Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004231.1 | NP_001004231.1 | 8 - 9 | 967 | 84 | |
XM_006233584.2 | XP_006233646.1 | 7 - 8 | 1605 | 84 | ||
GenBank mRNA | BC079156.1 | - | 8 - 9 | 967 | 7128989 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |