Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107818.1 | NP_001101288.1 | 7 - 8 | 914 | 101 | |
XM_006233612.2 | XP_006233674.1 | 6 - 7 | 1206 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |