Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014178.2 | NP_001014200.2 | 3 - 4 | 380 | 79 | |
GenBank mRNA | AI070071.1 | - | 3 - 4 | 380 | 79 | |
EV771144.1 | - | 1 - 2 | 60 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |