Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retnlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024281.1 | NP_001019452.1 | 2 - 3 | 179 | 83 | |
XM_008768646.1 | XP_008766868.1 | 1 - 2 | 248 | 83 | ||
GenBank mRNA | AY641429.1 | - | 2 - 3 | 179 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |