Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147139.2 | NP_671480.2 | 3 - 4 | 407 | 64 | |
GenBank mRNA | CB731093.1 | - | 3 - 4 | 377 | 64 | |
EV772196.1 | - | 2 - 3 | 260 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |