Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107922.2 | NP_001101392.2 | 3 - 4 | 952 | 115 | |
XM_006237989.2 | XP_006238051.1 | 3 - 4 | 1325 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |