Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_232840.8 | XP_232840.4 | 4 - 5 | 729 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |