Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106639.1 | NP_001100109.1 | 2 - 3 | 449 | 149 | |
XM_006237914.2 | XP_006237976.1 | 2 - 3 | 479 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |